Assay Details
| Gene Symbol: Sgk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037759.1 | NP_001032848.1 | 15 - 16 | 1461 | 128 | |
| NM_133220.2 | NP_573483.1 | 16 - 17 | 1580 | 128 | ||
| NM_177547.3 | NP_808215.2 | 15 - 16 | 1473 | 128 | ||
| GenBank mRNA | AF312007.1 | - | 16 - 17 | 1522 | 128 | |
| AK028722.1 | - | 15 - 16 | 1816 | 128 | ||
| AK030314.1 | - | 15 - 16 | 1462 | 128 | ||
| AK031133.1 | - | 16 - 17 | 1561 | 128 | ||
| AK031328.1 | - | 15 - 16 | 1402 | 128 | ||
| AK143836.1 | - | 16 - 17 | 1627 | 128 | ||
| AK146561.1 | - | 15 - 16 | 1416 | 128 | ||
| AK151181.1 | - | 16 - 17 | 1525 | 128 | ||
| AK151867.1 | - | 16 - 17 | 1557 | 128 | ||
| AK151996.1 | - | 16 - 17 | 1724 | 128 | ||
| AK166414.1 | - | 15 - 16 | 1404 | 128 | ||
| AK166531.1 | - | 15 - 16 | 1422 | 128 | ||
| AK171120.1 | - | 15 - 16 | 1449 | 128 | ||
| AK171186.1 | - | 15 - 16 | 1449 | 128 | ||
| AK171678.1 | - | 15 - 16 | 1433 | 128 | ||
| AK171959.1 | - | 15 - 16 | 1510 | 128 | ||
| AK172323.1 | - | 15 - 16 | 1423 | 128 | ||
| BC018363.1 | - | 13 - 14 | 1111 | 2631682 | 128 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sgk3
Genomic Map