Assay Details
| Gene Symbol: Sgk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037759.1 | NP_001032848.1 | 4 - 5 | 471 | 115 | |
| NM_133220.2 | NP_573483.1 | 5 - 6 | 590 | 115 | ||
| NM_177547.3 | NP_808215.2 | 4 - 5 | 483 | 115 | ||
| GenBank mRNA | AF312007.1 | - | 5 - 6 | 532 | 115 | |
| AK028722.1 | - | 5 - 6 | 535 | 115 | ||
| AK030314.1 | - | 4 - 5 | 472 | 115 | ||
| AK031133.1 | - | 5 - 6 | 571 | 115 | ||
| AK031328.1 | - | 4 - 5 | 412 | 115 | ||
| AK143836.1 | - | 5 - 6 | 578 | 115 | ||
| AK146561.1 | - | 4 - 5 | 426 | 115 | ||
| AK151181.1 | - | 5 - 6 | 535 | 115 | ||
| AK151793.1 | - | 4 - 5 | 463 | 115 | ||
| AK151867.1 | - | 5 - 6 | 567 | 115 | ||
| AK151996.1 | - | 5 - 6 | 734 | 115 | ||
| AK166414.1 | - | 4 - 5 | 412 | 115 | ||
| AK166531.1 | - | 4 - 5 | 433 | 115 | ||
| AK171120.1 | - | 4 - 5 | 459 | 115 | ||
| AK171186.1 | - | 4 - 5 | 459 | 115 | ||
| AK171678.1 | - | 4 - 5 | 442 | 115 | ||
| AK171959.1 | - | 4 - 5 | 520 | 115 | ||
| AK172323.1 | - | 4 - 5 | 433 | 115 | ||
| BC018363.1 | - | 2 - 3 | 121 | 2631682 | 115 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sgk3
Genomic Map