Assay Details
| Gene Symbol: Sgk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037759.1 | NP_001032848.1 | 7 - 8 | 695 | 110 | |
| NM_133220.2 | NP_573483.1 | 8 - 9 | 814 | 110 | ||
| NM_177547.3 | NP_808215.2 | 7 - 8 | 707 | 110 | ||
| GenBank mRNA | AF312007.1 | - | 8 - 9 | 756 | 110 | |
| AK028722.1 | - | 8 - 9 | 759 | 110 | ||
| AK030314.1 | - | 7 - 8 | 696 | 110 | ||
| AK031133.1 | - | 8 - 9 | 795 | 110 | ||
| AK031328.1 | - | 7 - 8 | 636 | 110 | ||
| AK143836.1 | - | 8 - 9 | 802 | 110 | ||
| AK146561.1 | - | 7 - 8 | 650 | 110 | ||
| AK151181.1 | - | 8 - 9 | 759 | 110 | ||
| AK151793.1 | - | 7 - 8 | 687 | 110 | ||
| AK151867.1 | - | 8 - 9 | 791 | 110 | ||
| AK151996.1 | - | 8 - 9 | 958 | 110 | ||
| AK166531.1 | - | 7 - 8 | 657 | 110 | ||
| AK171120.1 | - | 7 - 8 | 683 | 110 | ||
| AK171186.1 | - | 7 - 8 | 683 | 110 | ||
| AK171678.1 | - | 7 - 8 | 666 | 111 | ||
| AK171959.1 | - | 7 - 8 | 744 | 110 | ||
| AK172323.1 | - | 7 - 8 | 657 | 110 | ||
| BC018363.1 | - | 5 - 6 | 345 | 2631682 | 110 | |
More Information
Set Membership: |
|

Sgk3
Genomic Map