Assay Details
| Gene Symbol: Sgk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037759.1 | NP_001032848.1 | 8 - 9 | 752 | 90 | |
| NM_133220.2 | NP_573483.1 | 9 - 10 | 871 | 90 | ||
| NM_177547.3 | NP_808215.2 | 8 - 9 | 764 | 90 | ||
| GenBank mRNA | AF312007.1 | - | 9 - 10 | 813 | 90 | |
| AK028722.1 | - | 9 - 10 | 816 | 90 | ||
| AK030314.1 | - | 8 - 9 | 753 | 90 | ||
| AK031133.1 | - | 9 - 10 | 852 | 90 | ||
| AK031328.1 | - | 8 - 9 | 693 | 90 | ||
| AK143836.1 | - | 9 - 10 | 859 | 90 | ||
| AK146561.1 | - | 8 - 9 | 707 | 90 | ||
| AK151181.1 | - | 9 - 10 | 816 | 90 | ||
| AK151793.1 | - | 8 - 9 | 744 | 90 | ||
| AK151867.1 | - | 9 - 10 | 848 | 90 | ||
| AK151996.1 | - | 9 - 10 | 1015 | 90 | ||
| AK166531.1 | - | 8 - 9 | 714 | 90 | ||
| AK171120.1 | - | 8 - 9 | 740 | 90 | ||
| AK171186.1 | - | 8 - 9 | 740 | 90 | ||
| AK171678.1 | - | 8 - 9 | 91 | |||
| AK171959.1 | - | 8 - 9 | 801 | 90 | ||
| AK172323.1 | - | 8 - 9 | 714 | 90 | ||
| BC018363.1 | - | 6 - 7 | 402 | 2631682 | 90 | |
More Information
Set Membership: |
|

Sgk3
Genomic Map