Assay Details
| Gene Symbol: Cdc42ep2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026772.2 | NP_081048.1 | 1 - 2 | 56 | 117 | |
| GenBank mRNA | AK083004.1 | - | 57 | 117 | ||
| BB842420.1 | - | 1 - 2 | 57 | 117 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_026772.2 transcripts.
Set Membership: |
|

Cdc42ep2
Genomic Map