Assay Details
| Gene Symbol: Pawr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_054056.2 | NP_473397.1 | 2 - 3 | 683 | 66 | |
| GenBank mRNA | AF377871.1 | - | 1 - 2 | 86 | 66 | |
| AW123400.1 | - | 1 - 2 | 286 | 66 | ||
| DQ449074.1 | - | 1 - 2 | 494 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pawr
Genomic Map