Assay Details
| Gene Symbol: Pawr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_054056.2 | NP_473397.1 | 5 - 6 | 996 | 100 | |
| GenBank mRNA | AF377871.1 | - | 4 - 5 | 399 | 100 | |
| DQ363525.1 | - | 5 - 6 | 996 | 100 | ||
| DQ449073.1 | - | 3 - 4 | 675 | 100 | ||
| DQ449074.1 | - | 4 - 5 | 807 | 100 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pawr
Genomic Map