Assay Details
| Gene Symbol: Rgs20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177795.1 | NP_001171266.1 | 4 - 5 | 807 | 77 | |
| NM_001290372.1 | NP_001277301.1 | 3 - 4 | 390 | 77 | ||
| NM_021374.5 | NP_067349.2 | 3 - 4 | 460 | 77 | ||
| GenBank mRNA | AF191552.1 | - | 3 - 4 | 350 | 77 | |
| AF191554.1 | - | 3 - 4 | 336 | 77 | ||
| AK013773.1 | - | 3 - 4 | 289 | 77 | ||
| AK078208.1 | - | 3 - 4 | 305 | 77 | ||
| AK089947.1 | - | 3 - 4 | 280 | 77 | ||
| AK158864.1 | - | 3 - 4 | 301 | 77 | ||
| BC116925.1 | - | 3 - 4 | 350 | 40126244 | 77 | |
| BC116929.1 | - | 3 - 4 | 350 | 40126248 | 77 | |
| BI735670.1 | - | 4 - 5 | 570 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rgs20
Genomic Map