Assay Details
| Gene Symbol: Sox14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011440.1 | NP_035570.1 | 1 | 1945 | 135 | |
| GenBank mRNA | AK138208.1 | - | 1 | 1837 | 135 | |
| BC055767.1 | - | 1 | 1935 | 135 | ||
| BC069948.1 | - | 1 | 1835 | 135 | ||
| BC083360.1 | - | 1 | 1942 | 135 | ||
| BC100555.1 | - | 1 | 1946 | 135 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
visual perception
entrainment of circadian clock
regulation of neuron migration
RNA polymerase II transcription factor activity, sequence-specific DNA binding
DNA binding
chromatin binding
protein heterodimerization activity
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
visual perception
entrainment of circadian clock
regulation of neuron migration
RNA polymerase II transcription factor activity, sequence-specific DNA binding
DNA binding
chromatin binding
protein heterodimerization activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P641132 | BC083360.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P641132 | BC100555.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P641132 | BC055767.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P641132 | BC069948.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P641132 | AF193435.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P641132 | BC083360.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P641132 | BC100555.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P641132 | BC055767.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P641132 | BC069948.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P641132 | AF193435.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P117461 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P641132 | BC083360.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P641132 | BC100555.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P641132 | BC055767.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P641132 | BC069948.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P641132 | AF193435.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P641132 | BC083360.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P641132 | BC100555.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P641132 | BC055767.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P641132 | BC069948.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P641132 | AF193435.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P117461 | - | - |
| Whole Mouse Genome G4122A | A_51_P241897 | BC083360.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P241897 | BC100555.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P241897 | BC055767.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P241897 | BC069948.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P241897 | BC044735.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P641132 | BC083360.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P641132 | BC100555.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P641132 | BC055767.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P641132 | BC069948.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P641132 | AF193435.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P641132 | BC083360.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P641132 | BC100555.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P641132 | BC055767.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P641132 | BC069948.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P641132 | AF193435.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P117461 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07531335 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26766679 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26766666 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26766669 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26766649 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07788672 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26766647 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07531338 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26766654 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07531328 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26766656 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26766655 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26766671 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26766642 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26766645 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07788681 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07788662 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26766641 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26766657 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26766678 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26766643 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26766660 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26766675 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26766662 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26766661 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26766651 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10596025 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10596026 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10596024 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1427609_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10596023 | - | - |
| Mouse Genome 430 2.0 Array | 1427609_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1427609_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 20669_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 20669_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0900024147.mm.1 | NM_011440.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900024147.mm.1 | NM_011440.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900024147.mm.1 | TC0900002928.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900024145.mm.1 | NM_011440.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900024145.mm.1 | NM_011440.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900024145.mm.1 | TC0900002928.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900024146.mm.1 | NM_011440.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900024146.mm.1 | NM_011440.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900024146.mm.1 | TC0900002928.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900024143.mm.1 | NM_011440.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900024143.mm.1 | NM_011440.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900024143.mm.1 | TC0900002928.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900024144.mm.1 | NM_011440.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900024144.mm.1 | NM_011440.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900024144.mm.1 | TC0900002928.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1427609_PM_at | - | - |
| Mouse Expression Set 430 | 1427609_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427609_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427609_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427609_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427609_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 829890 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1257039 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2664638 | - | - |

Sox14
Genomic Map