Assay Details
| Gene Symbol: Taf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290729.1 | NP_001277658.1 | 19 - 20 | 3083 | 81 | |
| GenBank mRNA | AB299229.1 | - | 19 - 20 | 3010 | 81 | |
| AF022178.1 | - | 4 - 5 | 398 | 81 | ||
| AK050691.1 | - | 4 - 5 | 454 | 81 | ||
| BC047418.1 | - | 9 - 10 | 1196 | 81 | ||
| BC051073.1 | - | 6 - 7 | 809 | 81 | ||
| BC094568.1 | - | 1 - 2 | 367 | 81 | ||
More Information
Set Membership: |
|

Taf1
Genomic Map