Assay Details
| Gene Symbol: Taf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290729.1 | NP_001277658.1 | 31 - 32 | 4820 | 133 | |
| GenBank mRNA | AB299229.1 | - | 31 - 32 | 4747 | 133 | |
| AK046668.1 | - | 10 - 11 | 1309 | 133 | ||
| BC047418.1 | - | 21 - 22 | 2933 | 133 | ||
| BC094568.1 | - | 13 - 14 | 2104 | 133 | ||
More Information
Set Membership: |
|

Taf1
Genomic Map