Assay Details
| Gene Symbol: Taf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290729.1 | NP_001277658.1 | 9 - 10 | 1682 | 154 | |
| GenBank mRNA | AB299229.1 | - | 9 - 10 | 1609 | 154 | |
More Information
Set Membership: |
|

Taf1
Genomic Map