Assay Details
| Gene Symbol: Snx12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110310.1 | NP_001103780.1 | 2 - 3 | 390 | 78 | |
| NM_001110311.1 | NP_001103781.1 | 2 - 3 | 390 | 78 | ||
| NM_018875.2 | NP_061363.2 | 2 - 3 | 390 | 78 | ||
| GenBank mRNA | AF062484.1 | - | 2 - 3 | 317 | 78 | |
| AK011257.1 | - | 2 - 3 | 281 | 78 | ||
| AK049561.1 | - | 2 - 3 | 283 | 78 | ||
| AK131838.1 | - | 2 - 3 | 380 | 78 | ||
| AK146833.1 | - | 2 - 3 | 310 | 78 | ||
| AK148235.1 | - | 2 - 3 | 343 | 78 | ||
| AK168604.1 | - | 2 - 3 | 281 | 78 | ||
| AK170403.1 | - | 2 - 3 | 281 | 78 | ||
| AK170992.1 | - | 2 - 3 | 377 | 78 | ||
| BC002009.1 | - | 2 - 3 | 319 | 3592627 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Snx12
Genomic Map