Assay Details
| Gene Symbol: Gprc5c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110337.1 | NP_001103807.1 | 4 - 5 | 1682 | 79 | |
| GenBank mRNA | AK128950.1 | - | 4 - 5 | 1372 | 79 | |
| CO425185.1 | - | 2 - 3 | 250 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gprc5c
Genomic Map