Assay Details
| Gene Symbol: Uba7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023738.4 | NP_076227.1 | 10 - 11 | 1313 | 56 | |
| GenBank mRNA | AF288694.1 | - | 5 - 6 | 575 | 56 | |
| AK004894.2 | - | 10 - 11 | 1314 | 56 | ||
| AK089235.1 | - | 10 - 11 | 1317 | 56 | ||
| AK089256.1 | - | 4 - 5 | 745 | 56 | ||
| AK170421.1 | - | 10 - 11 | 1306 | 56 | ||
| AK170644.1 | - | 9 - 10 | 1688 | 56 | ||
| AK172467.1 | - | 9 - 10 | 1651 | 56 | ||
| BC037693.1 | - | 8 - 9 | 1087 | 56 | ||
| BC138843.1 | - | 10 - 11 | 1230 | 8861865 | 56 | |
More Information
Updated Mapping Information
Set Membership: |
|

Uba7
Genomic Map