Assay Details
| Gene Symbol: Uba7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023738.4 | NP_076227.1 | 13 - 14 | 1711 | 72 | |
| GenBank mRNA | AF288694.1 | - | 8 - 9 | 973 | 72 | |
| AK004894.2 | - | 13 - 14 | 1712 | 72 | ||
| AK054410.1 | - | 13 - 14 | 1718 | 72 | ||
| AK089235.1 | - | 13 - 14 | 1715 | 72 | ||
| AK129009.1 | - | 13 - 14 | 1621 | 72 | ||
| AK170421.1 | - | 13 - 14 | 1704 | 72 | ||
| AK170644.1 | - | 12 - 13 | 2086 | 72 | ||
| AK172467.1 | - | 12 - 13 | 2049 | 72 | ||
| BC037693.1 | - | 11 - 12 | 1485 | 72 | ||
| BC138843.1 | - | 13 - 14 | 1628 | 8861865 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Uba7
Genomic Map