Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 5089 | Gm5089 |
6530402A20, C030005G22Rik, EG328479 |
Anti-sense | Chr. 14 - 122365679 - 122451146 on NCBI Build GRCm38 | |
| Downstream | zinc finger protein of the cerebellum 5 | Zic5 |
1700049L20Rik, Opr |
8.2kb | Anti-sense | Chr. 14 - 122459160 - 122465716 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map