Assay Details
| Gene Symbol: Prim2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008922.2 | NP_032948.1 | 6 - 7 | 741 | 91 | |
| GenBank mRNA | AK076095.1 | - | 6 - 7 | 723 | 91 | |
| AK077760.1 | - | 6 - 7 | 728 | 91 | ||
| AK153970.1 | - | 6 - 7 | 702 | 91 | ||
| AK168246.1 | - | 6 - 7 | 734 | 91 | ||
| BC019500.1 | - | 6 - 7 | 702 | 3602929 | 91 | |
| D13545.1 | - | 6 - 7 | 705 | 91 | ||
| D17385.1 | - | 6 - 7 | 685 | 91 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prim2
Genomic Map