Assay Details
| Gene Symbol: Sema3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042779.2 | NP_001036244.1 | 14 - 15 | 1761 | 85 | |
| NM_001291537.1 | NP_001278466.1 | 14 - 15 | 2025 | 85 | ||
| NM_001291538.1 | NP_001278467.1 | 14 - 15 | 2019 | 85 | ||
| NM_001291539.1 | NP_001278468.1 | 13 - 14 | 1702 | 85 | ||
| NM_009153.3 | NP_033179.2 | 12 - 13 | 1571 | 85 | ||
| NR_111986.1 | 12 - 13 | 1581 | 85 | |||
| GenBank mRNA | AK014333.1 | - | 12 - 13 | 1517 | 85 | |
| AK033597.1 | - | 13 - 14 | 1712 | 85 | ||
| AK037034.1 | - | 14 - 15 | 1750 | 85 | ||
| AK043634.1 | - | 13 - 14 | 1655 | 85 | ||
| AK048364.1 | - | 13 - 14 | 1983 | 85 | ||
| AK148232.1 | - | 14 - 15 | 1722 | 85 | ||
| BC090669.1 | - | 13 - 14 | 1682 | 30696359 | 85 | |
| BC150758.1 | - | 11 - 12 | 1417 | 9087669 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sema3b
Genomic Map