Assay Details
| Gene Symbol: Apeh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146226.2 | NP_666338.2 | 10 - 11 | 1029 | 89 | |
| GenBank mRNA | AK149713.1 | - | 10 - 11 | 1031 | 89 | |
| AK179741.1 | - | 1 - 2 | 132 | 89 | ||
| BC005716.1 | - | 9 - 10 | 1155 | 89 | ||
| BC025494.1 | - | 10 - 11 | 1011 | 5310043 | 89 | |
| BC026594.1 | - | 5 - 6 | 562 | 89 | ||
More Information
Set Membership: |
|

Apeh
Genomic Map