Assay Details
| Gene Symbol: Apeh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146226.2 | NP_666338.2 | 2 - 3 | 174 | 71 | |
| GenBank mRNA | AK041271.1 | - | 2 - 3 | 171 | 71 | |
| AK149671.1 | - | 2 - 3 | 176 | 71 | ||
| AK149713.1 | - | 2 - 3 | 176 | 71 | ||
| BC005716.1 | - | 2 - 3 | 160 | 71 | ||
| BC025494.1 | - | 2 - 3 | 156 | 5310043 | 71 | |
| CJ134183.1 | - | 2 - 3 | 165 | 71 | ||
More Information
Set Membership: |
|

Apeh
Genomic Map