Assay Details
| Gene Symbol: Slc38a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199217.1 | NP_001186146.1 | 2 - 3 | 251 | 58 | |
| NM_001199218.1 | NP_001186147.1 | 2 - 3 | 271 | 58 | ||
| NM_023805.3 | NP_076294.2 | 2 - 3 | 419 | 58 | ||
| GenBank mRNA | AF159856.1 | - | 2 - 3 | 221 | 58 | |
| AK002607.1 | - | 2 - 3 | 237 | 58 | ||
| AK035155.1 | - | 439 | 58 | |||
| BC010678.1 | - | 2 - 3 | 251 | 58 | ||
| BC021324.1 | - | 2 - 3 | 305 | 58 | ||
| BC054846.1 | - | 2 - 3 | 394 | 6493334 | 58 | |
| BC055339.1 | - | 2 - 3 | 271 | 6415115 | 58 | |
| CJ183778.1 | - | 2 - 3 | 278 | 58 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc38a3
Genomic Map