Assay Details
| Gene Symbol: Slc38a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199217.1 | NP_001186146.1 | 4 - 5 | 440 | 63 | |
| NM_001199218.1 | NP_001186147.1 | 4 - 5 | 460 | 63 | ||
| NM_023805.3 | NP_076294.2 | 4 - 5 | 608 | 63 | ||
| GenBank mRNA | AF159856.1 | - | 4 - 5 | 410 | 63 | |
| AK002607.1 | - | 4 - 5 | 426 | 63 | ||
| AK035155.1 | - | 628 | 63 | |||
| BC010678.1 | - | 4 - 5 | 440 | 63 | ||
| BC021324.1 | - | 4 - 5 | 494 | 63 | ||
| BC054846.1 | - | 4 - 5 | 583 | 6493334 | 63 | |
| BC055339.1 | - | 4 - 5 | 460 | 6415115 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc38a3
Genomic Map