Assay Details
| Gene Symbol: Slc38a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199217.1 | NP_001186146.1 | 5 - 6 | 519 | 62 | |
| NM_001199218.1 | NP_001186147.1 | 5 - 6 | 539 | 62 | ||
| NM_023805.3 | NP_076294.2 | 5 - 6 | 687 | 62 | ||
| GenBank mRNA | AK002607.1 | - | 5 - 6 | 505 | 62 | |
| BC010678.1 | - | 5 - 6 | 519 | 62 | ||
| BC021324.1 | - | 5 - 6 | 573 | 62 | ||
| BC054846.1 | - | 5 - 6 | 662 | 6493334 | 62 | |
| BC055339.1 | - | 5 - 6 | 539 | 6415115 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc38a3
Genomic Map