Assay Details
| Gene Symbol: Sema3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243072.1 | NP_001230001.1 | 12 - 13 | 1747 | 98 | |
| NM_001243073.1 | NP_001230002.1 | 12 - 13 | 1587 | 98 | ||
| NM_009152.4 | NP_033178.2 | 11 - 12 | 2015 | 98 | ||
| GenBank mRNA | AK140817.1 | - | 12 - 13 | 1588 | 98 | |
| BC057588.1 | - | 11 - 12 | 1662 | 6416990 | 98 | |
| BC090844.1 | - | 12 - 13 | 1747 | 30532393 | 98 | |
| D85028.2 | - | 11 - 12 | 2017 | 98 | ||
| L40484.1 | - | 8 - 9 | 1048 | 98 | ||
| L41541.1 | - | 11 - 12 | 1366 | 98 | ||
| X85993.1 | - | 11 - 12 | 1476 | 98 | ||
More Information
Set Membership: |
|

Sema3a
Genomic Map