Assay Details
| Gene Symbol: Sema3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243072.1 | NP_001230001.1 | 6 - 7 | 929 | 73 | |
| NM_001243073.1 | NP_001230002.1 | 6 - 7 | 769 | 73 | ||
| NM_009152.4 | NP_033178.2 | 5 - 6 | 1197 | 73 | ||
| GenBank mRNA | AK140817.1 | - | 6 - 7 | 770 | 73 | |
| AK164242.1 | - | 8 - 9 | 972 | 73 | ||
| BC057588.1 | - | 5 - 6 | 844 | 6416990 | 73 | |
| BC090844.1 | - | 6 - 7 | 929 | 30532393 | 73 | |
| D85028.2 | - | 5 - 6 | 1199 | 73 | ||
| L41541.1 | - | 5 - 6 | 548 | 73 | ||
| X85993.1 | - | 5 - 6 | 658 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sema3a
Genomic Map