Assay Details
| Gene Symbol: Itm2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008409.2 | NP_032435.2 | 2 - 3 | 387 | 67 | |
| GenBank mRNA | AK014024.1 | - | 2 - 3 | 155 | 67 | |
| AK051956.1 | - | 2 - 3 | 366 | 67 | ||
| AK135229.1 | - | 2 - 3 | 339 | 67 | ||
| AK167351.1 | - | 2 - 3 | 388 | 67 | ||
| BC031420.1 | - | 2 - 3 | 354 | 3668557 | 67 | |
| BQ568915.1 | - | 2 - 3 | 355 | 67 | ||
| L38971.1 | - | 2 - 3 | 390 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Itm2a
Genomic Map