Assay Details
| Gene Symbol: Ndufs8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271443.1 | NP_001258372.1 | 3 - 4 | 179 | 92 | |
| NM_001271444.1 | NP_001258373.1 | 3 - 4 | 419 | 92 | ||
| NM_144870.5 | NP_659119.2 | 3 - 4 | 422 | 92 | ||
| GenBank mRNA | AK135069.1 | - | 3 - 4 | 180 | 92 | |
| AK149674.1 | - | 3 - 4 | 380 | 92 | ||
| AK153182.1 | - | 3 - 4 | 366 | 92 | ||
| AK153900.1 | - | 3 - 4 | 156 | 92 | ||
| AK164901.1 | - | 3 - 4 | 156 | 92 | ||
| AK167907.1 | - | 3 - 4 | 157 | 92 | ||
| BC021616.1 | - | 3 - 4 | 140 | 5132866 | 92 | |
| BC086766.1 | - | 3 - 4 | 133 | 6820786 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ndufs8
Genomic Map