Assay Details
| Gene Symbol: Coro1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011778.2 | NP_035908.1 | 7 - 8 | 942 | 85 | |
| GenBank mRNA | AF143956.1 | - | 7 - 8 | 949 | 85 | |
| AK008947.1 | - | 3 - 4 | 382 | 85 | ||
| AK149639.1 | - | 7 - 8 | 952 | 85 | ||
| AK159329.1 | - | 5 - 6 | 636 | 85 | ||
| AK205268.1 | - | 2 - 3 | 146 | 85 | ||
| BC029326.1 | - | 7 - 8 | 901 | 85 | ||
| BC131944.1 | - | 7 - 8 | 889 | 40130221 | 85 | |
| BC132617.1 | - | 7 - 8 | 889 | 40130894 | 85 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Coro1b
Genomic Map