Assay Details
| Gene Symbol: Coro1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011778.2 | NP_035908.1 | 1 - 2 | 70 | 80 | |
| GenBank mRNA | AF143956.1 | - | 1 - 2 | 77 | 80 | |
| AK149639.1 | - | 1 - 2 | 80 | 80 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the AF143956.1,AK149639.1,NM_011778.2 transcripts.
This assay is in the 5'UTR region of the AF143956.1,AK149639.1,NM_011778.2 transcripts.
Set Membership: |
|

Coro1b
Genomic Map