Assay Details
| Gene Symbol: Coro1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011778.2 | NP_035908.1 | 2 - 3 | 280 | 58 | |
| GenBank mRNA | AF143956.1 | - | 2 - 3 | 287 | 58 | |
| AK149639.1 | - | 2 - 3 | 290 | 58 | ||
| AK209750.1 | - | 1 - 2 | 149 | 58 | ||
| AK213023.1 | - | 1 - 2 | 149 | 58 | ||
| BC029326.1 | - | 2 - 3 | 239 | 58 | ||
| BC131944.1 | - | 2 - 3 | 227 | 40130221 | 58 | |
| BC132617.1 | - | 2 - 3 | 227 | 40130894 | 58 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Coro1b
Genomic Map