Assay Details
| Gene Symbol: Pola2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164057.1 | NP_001157529.1 | 7 - 8 | 1044 | 74 | |
| NM_008893.3 | NP_032919.2 | 7 - 8 | 1044 | 74 | ||
| GenBank mRNA | AK087989.1 | - | 7 - 8 | 1040 | 74 | |
| AK133832.1 | - | 7 - 8 | 1030 | 74 | ||
| AK150099.1 | - | 7 - 8 | 986 | 74 | ||
| AK152974.1 | - | 7 - 8 | 1009 | 74 | ||
| AK153081.1 | - | 7 - 8 | 1011 | 74 | ||
| AK163733.1 | - | 7 - 8 | 982 | 74 | ||
| AK213846.1 | - | 2 - 3 | 194 | 74 | ||
| BC006945.1 | - | 7 - 8 | 990 | 3602559 | 74 | |
| BC021424.1 | - | 7 - 8 | 1019 | 5011204 | 74 | |
| BC023688.1 | - | 7 - 8 | 1058 | 5343583 | 74 | |
| BC064795.1 | - | 7 - 8 | 960 | 30111287 | 74 | |
| D13546.1 | - | 7 - 8 | 977 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pola2
Genomic Map