Assay Details
| Gene Symbol: Mus81 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027877.3 | NP_082153.3 | 6 - 7 | 822 | 133 | |
| GenBank mRNA | AF425647.1 | - | 6 - 7 | 637 | 133 | |
| AK145858.1 | - | 6 - 7 | 772 | 133 | ||
| BC026560.1 | - | 6 - 7 | 835 | 5038349 | 133 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mus81
Genomic Map