Assay Details
| Gene Symbol: Mus81 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027877.3 | NP_082153.3 | 9 - 10 | 1170 | 103 | |
| GenBank mRNA | AF425647.1 | - | 9 - 10 | 985 | 103 | |
| AK004647.1 | - | 7 - 8 | 1256 | 103 | ||
| AK145858.1 | - | 9 - 10 | 1120 | 103 | ||
| BC026560.1 | - | 9 - 10 | 1183 | 5038349 | 103 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mus81
Genomic Map