Assay Details
| Gene Symbol: Fibp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253832.1 | NP_001240761.1 | 4 - 5 | 595 | 76 | |
| NM_021438.4 | NP_067413.1 | 4 - 5 | 595 | 76 | ||
| GenBank mRNA | AF270700.1 | - | 4 - 5 | 508 | 76 | |
| AK008093.1 | - | 4 - 5 | 586 | 76 | ||
| AK012629.1 | - | 4 - 5 | 578 | 76 | ||
| AK028029.1 | - | 4 - 5 | 586 | 76 | ||
| AK086610.1 | - | 1 - 2 | 162 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fibp
Genomic Map