Assay Details
| Gene Symbol: Gpr137 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177361.1 | NP_001170832.1 | 1 - 2 | 530 | 126 | |
| GenBank mRNA | AK143450.1 | - | 1 - 2 | 531 | 126 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gpr137
Genomic Map