Assay Details
| Gene Symbol: Gpr137 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177360.1 | NP_001170831.1 | 2 - 3 | 865 | 67 | |
| NM_001177361.1 | NP_001170832.1 | 4 - 5 | 1038 | 67 | ||
| NM_001177362.1 | NP_001170833.1 | 4 - 5 | 729 | 67 | ||
| NM_207220.2 | NP_997103.1 | 4 - 5 | 1141 | 67 | ||
| GenBank mRNA | AK039612.1 | - | 4 - 5 | 641 | 67 | |
| AK142928.1 | - | 2 - 3 | 463 | 67 | ||
| AK143450.1 | - | 4 - 5 | 1039 | 67 | ||
| AK170308.1 | - | 4 - 5 | 731 | 67 | ||
| BC048078.1 | - | 5 - 6 | 1013 | 6308873 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gpr137
Genomic Map