Assay Details
| Gene Symbol: Gpr137 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177360.1 | NP_001170831.1 | 4 - 5 | 1108 | 103 | |
| NM_001177361.1 | NP_001170832.1 | 6 - 7 | 1281 | 103 | ||
| NM_001177362.1 | NP_001170833.1 | 6 - 7 | 972 | 103 | ||
| NM_207220.2 | NP_997103.1 | 6 - 7 | 1384 | 103 | ||
| GenBank mRNA | AK039612.1 | - | 6 - 7 | 884 | 103 | |
| AK142928.1 | - | 4 - 5 | 706 | 103 | ||
| AK143450.1 | - | 6 - 7 | 1282 | 103 | ||
| AK170308.1 | - | 6 - 7 | 974 | 103 | ||
| BC048078.1 | - | 7 - 8 | 1256 | 6308873 | 103 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31936820 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31936822 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31936819 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31936817 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31936818 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31936816 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2091974 | AK143450.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2091974 | AK170308.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2091974 | BC048078.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2091974 | NM_001177362.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2091974 | NM_207220.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 209449 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 763957 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P167704 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2091974 | AK143450.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2091974 | AK170308.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2091974 | BC048078.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2091974 | NM_001177362.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2091974 | NM_207220.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31936801 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31936793 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31936815 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31936814 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31936817 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31936799 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31936797 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31936805 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31936819 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31936810 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31936812 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31936806 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31936807 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31936804 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31936790 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31936788 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31936811 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31936795 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31936821 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P167704 | - | - |
| Whole Mouse Genome G4122A | A_52_P192651 | AY255566.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P192651 | BC048078.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P192651 | CX200961.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P192651 | NM_207220.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P192651 | AK142928.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P135276 | AK143450.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P135276 | AK170308.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P135276 | BC048078.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P135276 | NM_001177362.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P135276 | NM_207220.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P113051 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P167704 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14481246 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14232403 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14481245 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31936820 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31936822 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31936818 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31936816 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31936819 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31936817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31936763 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31936743 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31936790 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31936810 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31936762 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31936799 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31936812 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31936729 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31936817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31936814 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31936735 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31936774 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31936815 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31936766 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31936773 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31936768 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31936771 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31936807 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31936781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31936730 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31936797 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31936788 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31936772 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31936804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31936756 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31936770 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31936732 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31936819 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31936757 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31936821 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31936776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31936795 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31936801 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31936779 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31936754 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31936738 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31936767 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31936749 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31936793 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31936755 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31936817 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31936797 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14510042 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14232405 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31936801 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31936812 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31936793 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31936788 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31936795 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31936815 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14510033 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31936814 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31936805 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31936810 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31936804 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31936790 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10465519 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465515 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465514 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465509 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465517 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465512 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465518 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465510 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465511 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465513 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1436377_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10465508 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10461017 | - | - |
| Mouse Genome 430 2.0 Array | 1436377_at | - | - |
| Mouse Genome 430 2.0 Array | 1436377_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 107173_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 107173_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1900009495.mm.1 | NM_001177360.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900009495.mm.1 | NM_001177360.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900009495.mm.1 | NM_001177361.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900009495.mm.1 | NM_001177361.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900009495.mm.1 | NM_001177362.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900009495.mm.1 | NM_001177362.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900009495.mm.1 | NM_207220.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900009495.mm.1 | NM_207220.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900009495.mm.1 | TC1900001023.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1436377_at | - | - |
| Mouse Expression Set 430 | 1436377_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1436377_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1436377_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 637192 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2630347 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2630347 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002630347 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1219842 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1219842 | - | - |
| mouse-6 v1.1 (Illumina) | scl0107173.1_330-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2630347 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0107173.1_330-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2630347 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2780876 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2780876 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0107173.1_330-S | - | - |

Gpr137
Genomic Map