Assay Details
| Gene Symbol: Mras | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008624.3 | NP_032650.1 | 3 - 4 | 771 | 64 | |
| GenBank mRNA | AB004879.1 | - | 3 - 4 | 755 | 64 | |
| AF031159.1 | - | 2 - 3 | 351 | 64 | ||
| AF043581.1 | - | 2 - 3 | 369 | 64 | ||
| AK050469.1 | - | 3 - 4 | 615 | 64 | ||
| AK164066.1 | - | 3 - 4 | 415 | 64 | ||
| BC024389.1 | - | 3 - 4 | 488 | 4973582 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mras
Genomic Map