Assay Details
| Gene Symbol: Slc25a23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025877.4 | NP_080153.2 | 2 - 3 | 473 | 67 | |
| GenBank mRNA | AK206310.1 | - | 2 - 3 | 202 | 67 | |
| BC072660.1 | - | 2 - 3 | 469 | 30360302 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc25a23
Genomic Map