Assay Details
| Gene Symbol: Qrich1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175143.5 | NP_780352.2 | 1 - 2 | 158 | 122 | |
| GenBank mRNA | AK083212.1 | - | 1 - 2 | 143 | 122 | |
| BB851544.1 | - | 1 - 2 | 159 | 122 | ||
| BC006738.1 | - | 1 - 2 | 135 | 3156574 | 122 | |
| BC120916.1 | - | 1 - 2 | 89 | 122 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK083212.1, BC006738.1, NM_175143.5 transcripts.
Set Membership: |
|

Qrich1
Genomic Map