Assay Details
| Gene Symbol: Qrich1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114119.1 | NP_001107591.1 | 5 - 6 | 1823 | 111 | |
| NM_175143.5 | NP_780352.2 | 4 - 5 | 1699 | 111 | ||
| GenBank mRNA | AK083212.1 | - | 4 - 5 | 1684 | 111 | |
| AK151491.1 | - | 5 - 6 | 1797 | 111 | ||
| AK151530.1 | - | 5 - 6 | 1797 | 111 | ||
| BC006738.1 | - | 4 - 5 | 1676 | 3156574 | 111 | |
| BC120915.1 | - | 3 - 4 | 598 | 111 | ||
| BC120916.1 | - | 4 - 5 | 1630 | 111 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Qrich1
Genomic Map