Assay Details
| Gene Symbol: Qrich1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114119.1 | NP_001107591.1 | 6 - 7 | 1968 | 123 | |
| NM_175143.5 | NP_780352.2 | 5 - 6 | 1844 | 123 | ||
| GenBank mRNA | AK083212.1 | - | 5 - 6 | 1829 | 123 | |
| AK151491.1 | - | 6 - 7 | 1942 | 123 | ||
| AK151530.1 | - | 6 - 7 | 1942 | 123 | ||
| BC006738.1 | - | 5 - 6 | 1821 | 3156574 | 123 | |
| BC120915.1 | - | 4 - 5 | 743 | 123 | ||
| BC120916.1 | - | 5 - 6 | 1775 | 123 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Qrich1
Genomic Map