Assay Details
| Gene Symbol: Prkd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001171004.1 | NP_001164475.1 | 14 - 15 | 2649 | 74 | |
| NM_001171005.1 | NP_001164476.1 | 13 - 14 | 1845 | 74 | ||
| NM_029239.3 | NP_083515.2 | 14 - 15 | 2646 | 74 | ||
| GenBank mRNA | AK030058.1 | - | 13 - 14 | 1842 | 74 | |
| BC027320.1 | - | 13 - 14 | 2048 | 74 | ||
| BC037012.1 | - | 13 - 14 | 2290 | 5342428 | 74 | |
| BC089164.1 | - | 14 - 15 | 2548 | 5400250 | 74 | |
More Information
Set Membership: |
|

Prkd3
Genomic Map