Assay Details
| Gene Symbol: Trip10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001242389.1 | NP_001229318.1 | 12 - 13 | 1492 | 63 | |
| NM_001242390.1 | NP_001229319.1 | 12 - 13 | 1492 | 63 | ||
| NM_001242391.1 | NP_001229320.1 | 11 - 12 | 1324 | 63 | ||
| NM_134125.4 | NP_598886.1 | 11 - 12 | 1324 | 63 | ||
| GenBank mRNA | AF502565.1 | - | 12 - 13 | 1395 | 63 | |
| AK088909.1 | - | 11 - 12 | 1274 | 63 | ||
| AK149902.1 | - | 11 - 12 | 1296 | 63 | ||
| AY081142.1 | - | 12 - 13 | 1443 | 63 | ||
| BC003249.1 | - | 11 - 12 | 1326 | 3500118 | 63 | |
| CD348583.1 | - | 1 - 2 | 67 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Trip10
Genomic Map