Assay Details
| Gene Symbol: Kitl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013598.2 | NP_038626.1 | 6 - 7 | 806 | 72 | |
| GenBank mRNA | AK018777.1 | - | 6 - 7 | 806 | 72 | |
| AK134301.1 | - | 6 - 7 | 806 | 72 | ||
| AK165233.1 | - | 6 - 7 | 804 | 72 | ||
| M57647.1 | - | 6 - 7 | 758 | 72 | ||
| U44725.1 | - | 6 - 7 | 806 | 72 | ||
| X68989.1 | - | 5 - 6 | 72 | |||
| X95379.1 | - | 6 - 7 | 72 | |||
| X95380.1 | - | 6 - 7 | 72 | |||
| X95381.1 | - | 6 - 7 | 72 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Kitl
Genomic Map