Assay Details
| Gene Symbol: Kitl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013598.2 | NP_038626.1 | 8 - 9 | 980 | 111 | |
| GenBank mRNA | AK018777.1 | - | 8 - 9 | 980 | 111 | |
| AK134301.1 | - | 8 - 9 | 980 | 111 | ||
| AK165233.1 | - | 8 - 9 | 978 | 111 | ||
| BC011322.1 | - | 8 - 9 | 950 | 3662792 | 111 | |
| M57647.1 | - | 8 - 9 | 932 | 111 | ||
| S40364.1 | - | 8 - 9 | 888 | 111 | ||
| S40534.1 | - | 7 - 8 | 804 | 111 | ||
| U44725.1 | - | 8 - 9 | 980 | 111 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Kitl
Genomic Map