Assay Details
| Gene Symbol: Ralbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198949.1 | NP_001185878.1 | 2 - 3 | 542 | 99 | |
| NM_009067.5 | NP_033093.2 | 2 - 3 | 866 | 99 | ||
| GenBank mRNA | AK019116.1 | - | 2 - 3 | 421 | 99 | |
| AK133613.1 | - | 2 - 3 | 425 | 99 | ||
| AK136116.1 | - | 2 - 3 | 631 | 99 | ||
| AK138197.1 | - | 2 - 3 | 910 | 99 | ||
| AK159404.1 | - | 2 - 3 | 419 | 99 | ||
| BC054329.1 | - | 2 - 3 | 457 | 99 | ||
| BC067073.1 | - | 1 - 2 | 301 | 30611880 | 99 | |
| BC075732.1 | - | 2 - 3 | 491 | 6507517 | 99 | |
| BC076636.1 | - | 2 - 3 | 458 | 30658617 | 99 | |
| BC138546.1 | - | 1 - 2 | 307 | 8861566 | 99 | |
| BC145320.1 | - | 1 - 2 | 307 | 9053875 | 99 | |
| BC145815.1 | - | 1 - 2 | 307 | 40131118 | 99 | |
| X80937.1 | - | 2 - 3 | 443 | 99 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ralbp1
Genomic Map