Assay Details
| Gene Symbol: Ralbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009067.5 | NP_033093.2 | 1 - 2 | 568 | 76 | |
| GenBank mRNA | AK133613.1 | - | 1 - 2 | 127 | 76 | |
| AK136116.1 | - | 1 - 2 | 333 | 76 | ||
| AK138197.1 | - | 1 - 2 | 612 | 76 | ||
| AK145387.1 | - | 1 - 2 | 108 | 76 | ||
| BC028516.1 | - | 1 - 2 | 116 | 76 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK133613.1, AK136116.1, AK138197.1, AK145387.1, BC028516.1, NM_009067.5 transcripts.
Set Membership: |
|

Ralbp1
Genomic Map