Assay Details
| Gene Symbol: Ppm1m | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB474373.1 | - | 2 - 3 | 429 | 90 | |
| AK087999.1 | - | 2 - 3 | 428 | 90 | ||
| AY332616.1 | - | 2 - 3 | 397 | 90 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK087999.1 transcripts.
Set Membership: |
|

Ppm1m
Genomic Map